Assay Details
| Gene Symbol: moe-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064394.3 | NP_496795.1 | 7 - 8 | 1056 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

moe-3| Gene Symbol: moe-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064394.3 | NP_496795.1 | 7 - 8 | 1056 | 66 | |
Set Membership: |
|