Assay Details
| Gene Symbol: ztf-22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064410.4 | NP_496811.1 | 4 - 5 | 1230 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ztf-22| Gene Symbol: ztf-22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064410.4 | NP_496811.1 | 4 - 5 | 1230 | 65 | |
Set Membership: |
|