Assay Details
| Gene Symbol: ddl-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064446.4 | NP_496847.1 | 2 - 3 | 849 | 66 | |
| Gene Symbol: Y48E1B.17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383984.1 | 2 - 3 | 849 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383984.1 transcripts.
Set Membership: |
|

ddl-2
Genomic Map