Assay Details
| Gene Symbol: mrpl-37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064449.4 | NP_496850.3 | 3 - 4 | 546 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mrpl-37| Gene Symbol: mrpl-37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064449.4 | NP_496850.3 | 3 - 4 | 546 | 77 | |
Set Membership: |
|