Assay Details
| Gene Symbol: csp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027281.1 | NP_001022452.1 | 1 - 2 | 125 | 129 | |
| GenBank mRNA | AF088285.1 | - | 3 - 4 | 297 | 129 | |
More Information
Set Membership: |
|

csp-1
Genomic Map