Assay Details
| Gene Symbol: csp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027281.1 | NP_001022452.1 | 4 - 5 | 764 | 84 | |
| GenBank mRNA | AF088285.1 | - | 6 - 7 | 936 | 84 | |
More Information
Set Membership: |
|

csp-1
Genomic Map