Assay Details
| Gene Symbol: cup-14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267478.2 | NP_001254407.1 | 2 - 3 | 172 | 63 | |
| NM_001267479.3 | NP_001254408.1 | 1 - 2 | 142 | 63 | ||
More Information
Set Membership: |
|

cup-14
Genomic Map