Assay Details
| Gene Symbol: cup-14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267478.2 | NP_001254407.1 | 4 - 5 | 452 | 109 | |
| NM_001267479.3 | NP_001254408.1 | 3 - 4 | 422 | 109 | ||
More Information
Set Membership: |
|

cup-14
Genomic Map