Assay Details
| Gene Symbol: cup-14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267478.2 | NP_001254407.1 | 7 - 8 | 1601 | 82 | |
| NM_001267479.3 | NP_001254408.1 | 6 - 7 | 1571 | 82 | ||
More Information
Set Membership: |
|

cup-14
Genomic Map