Assay Details
| Gene Symbol: ing-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064508.5 | NP_496909.1 | 5 - 6 | 1376 | 105 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ing-3| Gene Symbol: ing-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064508.5 | NP_496909.1 | 5 - 6 | 1376 | 105 | |
Set Membership: |
|