Assay Details
| Gene Symbol: mcd-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267483.3 | NP_001254412.1 | 1 - 2 | 184 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mcd-1| Gene Symbol: mcd-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267483.3 | NP_001254412.1 | 1 - 2 | 184 | 63 | |
Set Membership: |
|