Assay Details
Gene Symbol: mcd-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267483.3 | NP_001254412.1 | 2 - 3 | 302 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
