Assay Details
| Gene Symbol: ace-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064562.5 | NP_496963.1 | 6 - 7 | 1598 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ace-3| Gene Symbol: ace-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064562.5 | NP_496963.1 | 6 - 7 | 1598 | 94 | |
Set Membership: |
|