Assay Details
| Gene Symbol: gcy-29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001377902.1 | NP_001364597.1 | 10 - 11 | 2079 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gcy-29| Gene Symbol: gcy-29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001377902.1 | NP_001364597.1 | 10 - 11 | 2079 | 71 | |
Set Membership: |
|