Assay Details
| Gene Symbol: tps-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001377904.2 | NP_001364816.1 | 3 - 4 | 474 | 75 | |
| NM_064634.6 | NP_497035.3 | 12 - 13 | 3427 | 75 | ||
| GenBank mRNA | AJ811574.1 | - | 12 - 13 | 3449 | 75 | |
| Gene Symbol: arl-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064636.3 | NP_497037.1 | 5 - 6 | 2321 | 75 | |
More Information
Set Membership: |
|

tps-2
Genomic Map