Assay Details
| Gene Symbol: tps-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001377904.2 | NP_001364816.1 | 4 - 5 | 601 | 146 | |
| NM_064634.6 | NP_497035.3 | 13 - 14 | 3554 | 146 | ||
| GenBank mRNA | AJ811574.1 | - | 13 - 14 | 3576 | 146 | |
| Gene Symbol: arl-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064636.3 | NP_497037.1 | 6 - 7 | 2448 | 146 | |
More Information
Set Membership: |
|

tps-2
Genomic Map