Assay Details
| Gene Symbol: tps-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064634.6 | NP_497035.3 | 1 - 2 | 344 | 73 | |
| GenBank mRNA | AJ811574.1 | - | 1 - 2 | 366 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tps-2| Gene Symbol: tps-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064634.6 | NP_497035.3 | 1 - 2 | 344 | 73 | |
| GenBank mRNA | AJ811574.1 | - | 1 - 2 | 366 | 73 | |
Set Membership: |
|