Assay Details
| Gene Symbol: tps-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064634.6 | NP_497035.3 | 2 - 3 | 460 | 90 | |
| GenBank mRNA | AJ811574.1 | - | 2 - 3 | 482 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tps-2| Gene Symbol: tps-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064634.6 | NP_497035.3 | 2 - 3 | 460 | 90 | |
| GenBank mRNA | AJ811574.1 | - | 2 - 3 | 482 | 90 | |
Set Membership: |
|