Assay Details
| Gene Symbol: tps-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064634.6 | NP_497035.3 | 3 - 4 | 623 | 100 | |
| GenBank mRNA | AJ811574.1 | - | 3 - 4 | 645 | 100 | |
More Information
Set Membership: |
|

tps-2
Genomic Map