Assay Details
| Gene Symbol: tps-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064634.6 | NP_497035.3 | 8 - 9 | 2772 | 108 | |
| GenBank mRNA | AJ811574.1 | - | 8 - 9 | 2794 | 108 | |
| Gene Symbol: arl-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064636.3 | NP_497037.1 | 2 - 3 | 1621 | 108 | |
More Information
Set Membership: |
|

tps-2
Genomic Map