Assay Details
| Gene Symbol: arl-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064636.3 | NP_497037.1 | 1 - 2 | 1161 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

arl-3| Gene Symbol: arl-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064636.3 | NP_497037.1 | 1 - 2 | 1161 | 62 | |
Set Membership: |
|