Assay Details
| Gene Symbol: mltn-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064637.7 | NP_497038.2 | 9 - 10 | 1997 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mltn-9| Gene Symbol: mltn-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064637.7 | NP_497038.2 | 9 - 10 | 1997 | 86 | |
Set Membership: |
|