Assay Details
| Gene Symbol: nsun-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306619.3 | NP_001293548.1 | 1 - 2 | 216 | 85 | |
| NM_001381671.1 | NP_001366974.1 | 2 - 3 | 408 | 85 | ||
| NM_001381672.1 | NP_001366973.1 | 2 - 3 | 439 | 85 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001381672.1 transcripts.
Set Membership: |
|

nsun-5
Genomic Map