Assay Details
| Gene Symbol: nsun-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306619.3 | NP_001293548.1 | 3 - 4 | 587 | 59 | |
| NM_001381671.1 | NP_001366974.1 | 4 - 5 | 779 | 59 | ||
| NM_001381672.1 | NP_001366973.1 | 4 - 5 | 810 | 59 | ||
More Information
Set Membership: |
|

nsun-5
Genomic Map