Assay Details
| Gene Symbol: trip-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064699.5 | NP_497100.1 | 4 - 5 | 1031 | 82 | |
| GenBank mRNA | AF197575.1 | - | 4 - 5 | 1041 | 82 | |
More Information
Set Membership: |
|

trip-4
Genomic Map