Assay Details
| Gene Symbol: ptr-20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064713.5 | NP_497114.1 | 8 - 9 | 1454 | 105 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ptr-20| Gene Symbol: ptr-20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064713.5 | NP_497114.1 | 8 - 9 | 1454 | 105 | |
Set Membership: |
|