Assay Details
| Gene Symbol: fbxb-81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064753.4 | NP_497154.2 | 1 - 2 | 84 | 121 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

fbxb-81| Gene Symbol: fbxb-81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064753.4 | NP_497154.2 | 1 - 2 | 84 | 121 | |
Set Membership: |
|