Assay Details
| Gene Symbol: F40G9.17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381685.1 | NP_001367841.1 | 3 - 4 | 394 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

F40G9.17| Gene Symbol: F40G9.17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381685.1 | NP_001367841.1 | 3 - 4 | 394 | 60 | |
Set Membership: |
|