Assay Details
| Gene Symbol: klp-20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064777.6 | NP_497178.1 | 10 - 11 | 1361 | 70 | |
| Gene Symbol: F40G9.17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381685.1 | NP_001367841.1 | 12 - 13 | 1727 | 70 | |
| Gene Symbol: hpo-38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001384002.1 | 12 - 13 | 1721 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001384002.1 transcripts.
Set Membership: |
|

klp-20
Genomic Map