Assay Details
Gene Symbol: F40G9.17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381685.1 | NP_001367841.1 | 21 - 22 | 3119 | 92 |
Gene Symbol: hpo-38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001384002.1 | 21 - 22 | 3113 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
