Assay Details
| Gene Symbol: helq-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027740.6 | NP_001022911.1 | 7 - 8 | 1470 | 66 | |
| NM_001373832.1 | NP_001359796.1 | 7 - 8 | 1412 | 66 | ||
More Information
Set Membership: |
|

helq-1
Genomic Map