Assay Details
| Gene Symbol: elf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026009.3 | NP_001021180.1 | 6 - 7 | 1242 | 136 | |
| NM_001026010.4 | NP_001021181.1 | 8 - 9 | 1499 | 136 | ||
More Information
Set Membership: |
|

elf-1
Genomic Map