Assay Details
| Gene Symbol: flp-24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064842.8 | NP_497243.3 | 1 - 2 | 217 | 79 | |
| GenBank mRNA | AY821517.1 | - | 1 - 2 | 142 | 79 | |
More Information
Set Membership: |
|

flp-24
Genomic Map