Assay Details
| Gene Symbol: pat-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381698.1 | NP_001367550.1 | 6 - 7 | 645 | 156 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pat-12| Gene Symbol: pat-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381698.1 | NP_001367550.1 | 6 - 7 | 645 | 156 | |
Set Membership: |
|