Assay Details
| Gene Symbol: cyn-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064856.7 | NP_497257.1 | 1 - 2 | 197 | 79 | |
| GenBank mRNA | U27354.1 | - | 1 - 2 | 197 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cyn-6| Gene Symbol: cyn-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064856.7 | NP_497257.1 | 1 - 2 | 197 | 79 | |
| GenBank mRNA | U27354.1 | - | 1 - 2 | 197 | 79 | |
Set Membership: |
|