Assay Details
| Gene Symbol: dac-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025958.4 | NP_001021129.1 | 6 - 7 | 862 | 89 | |
| NM_001025959.5 | NP_001021130.1 | 6 - 7 | 779 | 89 | ||
More Information
Set Membership: |
|

dac-1
Genomic Map