Assay Details
| Gene Symbol: lgc-46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064937.7 | NP_497338.2 | 10 - 11 | 1381 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

lgc-46| Gene Symbol: lgc-46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064937.7 | NP_497338.2 | 10 - 11 | 1381 | 84 | |
Set Membership: |
|