Assay Details
| Gene Symbol: cdh-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373846.2 | NP_001360600.1 | 23 - 24 | 8512 | 85 | |
| NM_001373847.2 | NP_001360601.1 | 16 - 17 | 5809 | 85 | ||
More Information
Set Membership: |
|

cdh-12
Genomic Map