Assay Details
| Gene Symbol: pals-17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065001.5 | NP_497402.2 | 5 - 6 | 593 | 165 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pals-17| Gene Symbol: pals-17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065001.5 | NP_497402.2 | 5 - 6 | 593 | 165 | |
Set Membership: |
|