Assay Details
| Gene Symbol: pals-18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027793.4 | NP_001022964.1 | 2 - 3 | 484 | 163 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pals-18| Gene Symbol: pals-18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027793.4 | NP_001022964.1 | 2 - 3 | 484 | 163 | |
Set Membership: |
|