Assay Details
| Gene Symbol: snf-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381736.1 | NP_001367976.1 | 4 - 5 | 487 | 119 | |
| GenBank mRNA | AY661556.1 | - | 4 - 5 | 428 | 119 | |
More Information
Set Membership: |
|

snf-6
Genomic Map