Assay Details
| Gene Symbol: polh-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065079.3 | NP_497480.2 | 1 - 2 | 147 | 57 | |
| GenBank mRNA | AB244413.1 | - | 1 - 2 | 130 | 57 | |
More Information
Set Membership: |
|

polh-1
Genomic Map