Assay Details
| Gene Symbol: polh-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065079.3 | NP_497480.2 | 4 - 5 | 659 | 72 | |
| GenBank mRNA | AB244413.1 | - | 4 - 5 | 642 | 72 | |
More Information
Set Membership: |
|

polh-1
Genomic Map