Assay Details
| Gene Symbol: dpy-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381759.2 | NP_001367968.1 | 8 - 9 | 1757 | 119 | |
| NM_001384037.1 | 12 - 13 | 2766 | 119 | |||
More Information
Set Membership: |
|

dpy-1
Genomic Map