Assay Details
| Gene Symbol: dpy-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381759.2 | NP_001367968.1 | 10 - 11 | 2655 | 69 | |
| NM_001384037.1 | 14 - 15 | 3664 | 69 | |||
More Information
Set Membership: |
|

dpy-1
Genomic Map