Assay Details
| Gene Symbol: dpy-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381759.2 | NP_001367968.1 | 2 - 3 | 435 | 127 | |
| NM_001384037.1 | 6 - 7 | 1051 | 127 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dpy-1| Gene Symbol: dpy-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381759.2 | NP_001367968.1 | 2 - 3 | 435 | 127 | |
| NM_001384037.1 | 6 - 7 | 1051 | 127 | |||
Set Membership: |
|