Assay Details
| Gene Symbol: dpy-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381759.2 | NP_001367968.1 | 7 - 8 | 1695 | 89 | |
| NM_001384037.1 | 11 - 12 | 2704 | 89 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dpy-1| Gene Symbol: dpy-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381759.2 | NP_001367968.1 | 7 - 8 | 1695 | 89 | |
| NM_001384037.1 | 11 - 12 | 2704 | 89 | |||
Set Membership: |
|