Assay Details
| Gene Symbol: smg-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381779.2 | NP_001367726.1 | 8 - 9 | 1925 | 134 | |
| NM_065165.8 | NP_497566.3 | 8 - 9 | 1926 | 134 | ||
More Information
Set Membership: |
|

smg-6
Genomic Map