Assay Details
| Gene Symbol: Y54F10AM.8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065169.7 | NP_497570.1 | 7 - 8 | 1443 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Y54F10AM.8| Gene Symbol: Y54F10AM.8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065169.7 | NP_497570.1 | 7 - 8 | 1443 | 61 | |
Set Membership: |
|