Assay Details
| Gene Symbol: let-805 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027470.3 | NP_001022641.1 | 19 - 20 | 11815 | 83 | |
| NM_001306713.4 | NP_001293642.1 | 2 - 3 | 271 | 83 | ||
| GenBank mRNA | AF148953.1 | - | 19 - 20 | 11822 | 83 | |
More Information
Set Membership: |
|

let-805
Genomic Map